missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse Galectin-1 ELISA Kit (Colorimetric)

Product Code. 18604880 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18604880 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18604880 Supplier Novus Biologicals™ Supplier No. NBP267971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.78 to 50 ng/mL
Assay Sensitivity 0.47 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Biological Fluids
Specificity This kit recognizes Mouse GAL1 in samples. No significant cross-reactivity or interference between Mouse GAL1 and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 3956
Gene Symbol LGALS1
Quantity 1 Kit
Research Discipline Apoptosis, Biologically Active Proteins
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.