missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Furin Convertase (aa 12-27) Synthetic Peptide

Product Code. 11538881
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11538881 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11538881 Supplier Invitrogen™ Supplier No. PEP010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Mouse Ferin Convertase (residues 12-27) Synthetic Peptide, PEP-010, from Invitrogen.

PEP-010 is a 14 amino acid (amino acids 780-793) synthetic peptide whose sequence is from the carboxyl end of the mouse furin convertase protein. The sequence of this peptide is (amino to carboxy terminus): R-G-E-R-T-A-F-I-K-D-Q-S-A-LThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and furin convertase, catalog # PA1-062 and PA1.1-062. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Species Mouse
Purification Method purified
Common Name Mouse Furin Convertase (aa 12-27)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence R - G - E - R - T - A - F - I - K - D - Q - S - A - L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.