missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Frizzled-2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1307
This item is not returnable.
View return policy
Description
Frizzled-2 Polyclonal specifically detects Frizzled-2 in Mouse samples. It is validated for Western Blot.
Specifications
| Frizzled-2 | |
| Polyclonal | |
| Unconjugated | |
| Q9JIP6 | |
| FZD2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-2 Gln29-Leu168 Accession # Q9JIP6 | |
| 100 μg | |
| Primary | |
| Detects mouse Frizzled-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse (rm) Frizzled-1 is observed and less than 5% cross-reactivity with rmFrizzled-4, recombinant human Frizzled-5, rmFrizzled-6, and rmFrizzled-8 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| frizzled (Drosophila) homolog 2, frizzled homolog 2 (Drosophila), frizzled-2, Frizzled2, fz-2, Fz2, FZD2, FzE2, hFz2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2535 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction