missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FoxN1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB7295-SP
This item is not returnable.
View return policy
Description
FoxN1 Monoclonal specifically detects FoxN1 in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| FoxN1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q61575 | |
| FOXN1 | |
| E. coli-derived recombinant mouse FoxN1 Val2-Asp164 Accession # Q61575 | |
| 25 μg | |
| Primary | |
| Detects mouse FoxN1 in direct ELISAs. | |
| Mouse | |
| Purified |
| Immunohistochemistry | |
| 717008 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FKHL20, forkhead box N1, forkhead box protein N1, RONU, Rowett nude, WHN, WHNRONU, winged helix nude, winged-helix nude, Winged-helix transcription factor nude | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8456 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction