missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Flt3 Ligand, Recombinant Protein, Carrier-Free
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Flt3 (FMS-related tyrosine kinase 3) Ligand is a growth factor that stimulates the growth and differentiation of early hematopoetic progenitor cells. Flt3 Ligand alone exerts weak biological effects, and instead synergizes with other colony-stimulating factors to induce the proliferation and differentiation of cells of the myeloid and lymphoid lineages. Recent studies have suggested that Flt3 Ligand plays an important role in the development and expansion of dendritic cells in vivo, and that it may influence NK cell survival by this mechanism.
Flt3 Ligand transcripts are detectable in most cell types and tissues, although the protein has only been found in stromal cells and T cells. It is expressed most often as a transmembrane cell surface protein, although processing can yield a soluble, secreted isoform.
Flt3 Ligand transcripts are detectable in most cell types and tissues, although the protein has only been found in stromal cells and T cells. It is expressed most often as a transmembrane cell surface protein, although processing can yield a soluble, secreted isoform.
Specifications
Specifications
| Accession Number | NM_013520 |
| Concentration | 0.5mg/mL |
| For Use With (Application) | Cytokine Bioassay |
| Formulation | Sterile liquid: 20mM glycine, 150mM NaCl, pH 3.0; 0.22μm filtered. |
| Molecular Weight (g/mol) | 20.7kDa |
| Name | Flt3 Ligand |
| Purification Method | SDS-PAGE |
| Quantity | 100 μg |
| Source | E. coli |
| Storage Requirements | Store at less than or equal to -70°C. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction