missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Flt-3 Ligand (FLT3L) Recombinant Protein
Click to view available options
Quantity:
10 μg
100 μg
25 μg
Unit Size:
100µg
10µg
25µg
Description
FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
Specifications
Specifications
| Accession Number | P49772 |
| Concentration | 200 μg/mL |
| For Use With (Application) | Control, ELISA, Western Blot |
| Formulation | PBS with no preservative; pH 6 |
| Gene ID (Entrez) | 14256 |
| Molecular Weight (g/mol) | 18.3 kD |
| Name | Mouse FLT3LG (aa 27-188) |
| Quantity | 10 μg |
| Source | CHO cells |
| Storage Requirements | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction