missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Flt-3 Ligand (FLT3L), AOF Recombinant Protein

Product Code. 17676545
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 17676545

Brand: Invitrogen™ RP87733

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

Animal Origin Free Reconstitute with 0.1 mg/mL sterile water

FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
  • Purity: >95% by SDS-Page
  • Endotoxin Concentration: AX41 ≤EUs/μg
  • Protein Length: 163 aa.
  • Biological Activity: <10 ng/mL
  • Molecular Weight: 18.6 kDa
  • Animal Origin Free: Yes
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49772
For Use With (Application) Control, ELISA, Functional Assay, Gel Shift, Western Blot
Formulation 10 mM sodium phosphate with no preservative
Gene ID (Entrez) 14256
Molecular Weight (g/mol) 18.6 kDa
Name Mouse Flt3 Ligand (Flt3L), AOF
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias FL; Flt 3 ligand; Flt3 L; Flt3 ligand; Flt3l; FLT3LG; fms related tyrosine kinase 3 ligand; FMS-like tyrosine kinase 3 ligand; Fms-related tyrosine kinase 3 ligand; H-FLT-3-L; Ly72L; M-FLT-3-L; NEWGENE_2322792; SL cytokine
Gene Symbol Flt3l
Specific Reactivity Mouse
Conjugate Unconjugated
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.