missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fibroblast Activation Protein alpha/FAP Antibody, R&D Systems™

Rat Monoclonal Antibody
£166.00 - £441.00
Specifications
| Antigen | Fibroblast Activation Protein alpha/FAP |
|---|---|
| Clone | 983810 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713937
![]() |
R&D Systems
MAB97271-SP |
25 μg |
£166.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18740517
![]() |
R&D Systems
MAB97271-100 |
100 μg |
£441.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Mouse samples. It is validated for ELISA.Specifications
| Fibroblast Activation Protein alpha/FAP | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| P97321 | |
| 2191.0 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 983810 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
| Mouse myeloma cell line NS0-derived recombinant mouse Fibroblast Activation Protein alpha/FAP, Leu26-Asp761, Accession No. P97321 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Fibroblast Activation Protein alpha/FAP in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title