missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FGFR3 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB710-SP
This item is not returnable.
View return policy
Description
FGFR3 Monoclonal specifically detects FGFR3 in Mouse samples. It is validated for Neutralization.
Specifications
| FGFR3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2261 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization | |
| 136812 | |
| Neutralization 3-12 ug/mL | |
| NP_001156689 | |
| FGFR3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse FGF R3 Glu21-Tyr369 (Asn310Ile, Ala354 del) (predicted) Accession # NP_001156689 | |
| 25 μg | |
| Primary | |
| Detects mouse FGF R3 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with the (IIIb) and (IIIc) isoforms of recombinant human FGF R3 is observed. Neutralizes the bioactivity of mouse FGF R3. Functions equally as well in the neutralization of recombinant mouse (rm) FGF R3 alpha (IIIb) and rmFGF R3 alpha (IIIc). | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction