missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse FGF-basic (FGF-2/bFGF) Recombinant Protein
Recombinant Mouse Fibroblast Growth Factor Basic (also known as FGF Basic, bFGF, FGF 2, FGF-β or Heparin Binding Growth Factor) is a bioactive protein intended for use in cell culture applications
£192.00 - £6360.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 14173 |
Molecular Weight (g/mol) | 17 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | FGF-Basic |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10511865
|
Gibco™
PMG0034 |
10μg |
£192.00
Each |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10161153
|
Gibco™
PMG0035 |
25μg |
£372.00
Each |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10437253
|
Gibco™
PMG0031 |
100μg |
£1272.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10444373
|
Gibco™
PMG0033 |
1mg |
£6360.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mouse FGF-basic (FGF-2/bFGF) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
FGF-basic is involved in a number of biological processes including embryonic development and differentiation, neuron differentiation, survival, and regeneration, and the proliferation of cells of mesodermal origin and many cells of neuroectodermal, ectodermal, and endodermal origin.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of BALB/3T3 cells
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
17 kDa | |
FGF-Basic | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
E. coli | |
Recombinant, Ligand | |
FGFs (Fibroblast Growth Factors) | |
Gibco™ |
14173 | |
Sequential Chromatography | |
Mouse | |
Carrier-Free | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Microbiology, Angiogenesis, Oncology | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.