missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse FGF-9 Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP8637
Code : LE
Additional Details : Weight : 0.01000kg
Description
Fibroblast Growth Factor 9 (mouse) Recombinant Protein, RP-8637, from Invitrogen.
Fibroblast Growth Factor-9 (FGF-9) is a steroid-regulated mitogen and survival factor for nerve and mesenchymal cells. FGF-9 is an autocrine/paracrine growth factor considered to be important for the growth and survival of motorneurons and prostate tissue. Recombinant mouse G-CSF is a non-glycosylated protein, containing 179 amino acids, with a molecular weight of 19 kDa. Purity is typically greater than 95% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent proliferation of mouse M-NFS-60 cells and is typically less than 10-60 ng/mL. The specific activity of this protein is 1x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA.Specifications
P54130 | |
Control | |
14180 | |
Mouse FGF9 | |
10μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
M-FGF-9 | |
Fgf9 | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
0.1mg/mL | |
10mM sodium phosphate with no preservative; pH 7.5 | |
∽23.3 | |
7.5 | |
E. coli | |
RUO | |
Mouse FGF9 | |
Mouse | |
Ms | |
Recombinant | |
Recombinant |
in vitroresearch only