Learn More
Gibco™ Mouse FGF-7 (KGF) Recombinant Protein, PeproTech®
Description
450-60-1MG will be provided as 10 x 100 μg (450-60-100UG). 450-60-250UG will be provided as multiple 50 μg (450-60-50UG) and 100 μg (450-60-100UG) vials. 450-60-500UG will be provided as 5 x 100 μg (450-60-100UG). Recombinant Murine KGF (FGF-7) is an 18.8 kDa protein consisting of 164 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P36363 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 14178 |
| Molecular Weight (g/mol) | 18.8 |
| Name | Mouse FGF-7 (KGF) |
| Quantity | 5 x 100 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.