missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FGF-4 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5846
This item is not returnable.
View return policy
Description
FGF-4 Polyclonal specifically detects FGF-4 in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| FGF-4 | |
| Polyclonal | |
| Unconjugated | |
| P11403 | |
| FGF4 | |
| E. coli-derived recombinant mouse FGF-4 Ala30-Leu202 Accession # P11403 | |
| 100 μg | |
| Primary | |
| Detects mouse FGF-4 in direct ELISAs. In direct ELISAs, approximately 60% cross-reactivity with recombinant human FGF-4 is observed. | |
| Mouse | |
| IgG |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| FGF4, fibroblast growth factor 4, HBGF-4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2249 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction