missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fc gamma RIII (CD16) Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB19601N-025
This item is not returnable.
View return policy
Description
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Fc gamma RIII (CD16) | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2214 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 275003 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Q5D5J5 | |
| Fcgr3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIII Ala31-Thr215 Accession # Q5D5J5 | |
| 25 Tests | |
| Primary | |
| Detects mouse Fc gamma RIIIA/B (CD16) in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Fcrl3/CD16-2, rmFc gamma RIA or rhFc gamma RIIA is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction