missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fas Ligand/TNFSF6 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF526-SP
This item is not returnable.
View return policy
Description
Fas Ligand/TNFSF6 Polyclonal specifically detects Fas Ligand/TNFSF6 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| Fas Ligand/TNFSF6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 356 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.125-0.75 ug/mL | |
| Q544E9 | |
| Fasl | |
| Mouse myeloma cell line NS0-derived recombinant mouse Fas Ligand/TNFSF6 Pro132-Leu279 Accession # Q544E9 | |
| 25 μg | |
| Primary | |
| Detects mouse Fas Ligand/TNFSF6 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Fas Ligand is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction