missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Epiregulin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1068-SP
This item is not returnable.
View return policy
Description
Epiregulin Monoclonal specifically detects Epiregulin in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| Epiregulin | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.2-1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q61521 | |
| EREG | |
| E. coli-derived recombinant mouse Epiregulin Val56-Leu101 Accession # Q61521 | |
| 25 μg | |
| Primary | |
| Detects mouse Epiregulin in ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human Epiregulin is observed, but no cross-reactivity with recombinant mouse (rm) Epigen, recombinant human (rh) TGF-alpha , rhAmphiregulin, rmBetacellulin, rhEGF, rhHB-EGF, rhHRG-alpha , or rhHRG-beta is observed. In sandwich immunoassays, no cross-reactivity with rmAmphiregulin, rhNRG-1-beta 1, rmBetacellulin, rmEGF, rhHeregulin, rhHB-EGF, or rhTGF-alpha is observed. | |
| Mouse | |
| Purified |
| Neutralization, ELISA | |
| 189611 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| epiregulin, ER, EREG, proepiregulin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2069 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction