missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Ephrin-A4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF569
This item is not returnable.
View return policy
Description
Ephrin-A4 Polyclonal specifically detects Ephrin-A4 in Mouse samples. It is validated for Western Blot.
Specifications
| Ephrin-A4 | |
| Polyclonal | |
| Unconjugated | |
| O08542 | |
| EFNA4 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Ephrin-A4 Leu26-Gly176 Accession # O08542 | |
| 100 μg | |
| Primary | |
| Detects mouse Ephrin-A4 in direct ELISAs and Western blots. In Western blots, approximately 35% cross-reactivity with recombinant human (rh) Ephrin-A4 is observed and less than 5% cross-reactivity with rhEphrin-B3, recombinant mouse (rm) Ephrin-A1, rmEphrin-A2, rhEphrin-A5, and rhEphrin-A3 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| EFL4, EFL-4, EFNA4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EphrinA4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1945 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction