missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphB6 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1428-SP
This item is not returnable.
View return policy
Description
EphB6 Monoclonal specifically detects EphB6 in Mouse samples. It is validated for Western Blot.
Specifications
| EphB6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O08644 | |
| EPHB6 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphB6 Leu33-Ser587 Accession # O08644 | |
| 25 μg | |
| Primary | |
| Detects mouse EphB6 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human EphA1, recombinant mouse (rm) EphA2, rmEphA3, rmEphA4, recombinant rat (rr) EphA5, rmEphA6, rmEphA7, rmEphA8, rrEphB1, rmEphB2, rmEphB3, and rmEphB4 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 92224 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, Hep, HEPTyrosine-protein kinase-defective receptor EPH-6, Mep, MGC129910, MGC129911 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2051 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction