missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphB6 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1428
This item is not returnable.
View return policy
Description
EphB6 Monoclonal specifically detects EphB6 in Mouse samples. It is validated for Western Blot.
Specifications
| EphB6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O08644 | |
| EPHB6 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphB6 Leu33-Ser587 Accession # O08644 | |
| 500 μg | |
| Primary | |
| Detects mouse EphB6 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human EphA1, recombinant mouse (rm) EphA2, rmEphA3, rmEphA4, recombinant rat (rr) EphA5, rmEphA6, rmEphA7, rmEphA8, rrEphB1, rmEphB2, rmEphB3, and rmEphB4 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 92224 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, Hep, HEPTyrosine-protein kinase-defective receptor EPH-6, Mep, MGC129910, MGC129911 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2051 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering