missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphB4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF446-SP
This item is not returnable.
View return policy
Description
EphB4 Polyclonal specifically detects EphB4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| EphB4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, Htk, HTKephrin receptor EphB4, Mdk2, Myk1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2050 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
| P54761 | |
| EPHB4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphB4 Leu16-Ala539 Accession # P54761 | |
| 25 μg | |
| Primary | |
| Detects mouse EphB4 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) EphB6 and rmEphA3 is observed and less than 1% cross-reactivity with rmEphA2, rmEphA4, rmEphA6, rmEphA7, rmEphA8, rmEphB2, and rmEphB3 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction