missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphB2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marca: R&D Systems MAB4672-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
EphB2 Monoclonal specifically detects EphB2 in Mouse samples. It is validated for Western Blot.
Especificaciones
| EphB2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P54763 | |
| EPHB2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphB2 Val27-Lys548 Accession # P54763 | |
| 25 μg | |
| Primary | |
| Detects mouse EphB2 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human EphB2 is observed. In Western blots, no cross-reactivity with recombinant human EphB2 is observed. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) EphA1, 2, 3, 4, 5, 6, 7, 8, rmEphB4, 6, recombinant human EphA10, recombinant rat (rr) EphA10, or rrEphB1 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 512013 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAPB, Cek5, Drt, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, Erk, ERKHek5, HEK5, Nuk, PCBC, protein-tyrosine kinase HEK5, Qek2, Renal carcinoma antigen NY-REN-47, Sek3, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2048 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido