missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphA7 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB1495
This item is not returnable.
View return policy
Description
EphA7 Monoclonal specifically detects EphA7 in Mouse samples. It is validated for Western Blot.
Specifications
| EphA7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q61772 | |
| EPHA7 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA7 isoform 1 Ala30-Pro554 (Gly540-Glu544 del) Accession # Q61772 | |
| 500 μg | |
| Primary | |
| Detects mouse EphA7 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human EphA1, recombinant mouse EphA2, A3, A4, A6, A8, B2, B3, B4, recombinant rat (rr) EphA5, or rrEphB1. | |
| Mouse | |
| Purified |
| Western Blot | |
| 88712 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cek11, Ebk, EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, Mdk1, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2045 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction