missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphA3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF640
This item is not returnable.
View return policy
Description
EphA3 Polyclonal specifically detects EphA3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| EphA3 | |
| Polyclonal | |
| Unconjugated | |
| Q8BRB1 | |
| EPHA3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA3 Glu21-His541 (Thr323Ala, Glu476Gln) Accession # Q8BRB1 | |
| 100 μg | |
| Primary | |
| Detects mouse EphA3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) EphA4, rmEphA8, recombinant rat EphA5, and rmEphA6 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Cek4, EC 2.7.10, EC 2.7.10.1, EK4, EPH receptor A3, EphA3, EPH-like kinase 4, ephrin type-A receptor 3, ETK, hEK4, HEKETK1eph-like tyrosine kinase 1, human embryo kinase 1, Tyro4, TYRO4 protein tyrosine kinase, Tyrosine-protein kinase receptor ETK1, Tyrosine-protein kinase TYRO4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2042 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction