missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphA1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marke: R&D Systems MAB3034-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
EphA1 Monoclonal specifically detects EphA1 in Mouse samples. It is validated for Western Blot.
Spezifikation
| EphA1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q60750 | |
| EPHA1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA1 Glu27-Glu548 Accession # Q60750 | |
| 25 μg | |
| Primary | |
| Detects mouse EphA1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human EphA1 is observed and no cross-reactivity with recombinant mouse EphA3, A5, A6, A7, A8, B2, B3, B4, or B6 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 404515 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10, EC 2.7.10.1, EPH receptor A1, eph tyrosine kinase 1, EphA1, EPHephrin type-A receptor 1, EPHT, EPHT1erythropoietin-producing hepatoma amplified sequence, Esk, MGC163163, oncogene EPH, soluble EPHA1 variant 1, soluble EPHA1 variant 2, Tyrosine-protein kinase receptor EPH | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2041 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur