missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EPCR Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2749
This item is not returnable.
View return policy
Description
EPCR Polyclonal specifically detects EPCR in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry.
Specifications
| EPCR | |
| Polyclonal | |
| Biotin | |
| Q64695 | |
| PROCR | |
| Mouse myeloma cell line NS0-derived recombinant mouse EPCR Ala17-Ser214 Accession # Q64695 | |
| 50 μg | |
| Primary | |
| Detects mouse EPCR in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human EPCR is observed. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL | |
| APC receptor, CCCA, CCD41, CCD41centrosome-associated protein, CD201, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, PROCR, protein C receptor, endothelial | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10544 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction