missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EpCAM/TROP-1 Antibody, R&D Systems™
Rat Monoclonal Antibody
£206.00 - £486.00
Specifications
| Antigen | EpCAM/TROP1 |
|---|---|
| Clone | 1085639 |
| Dilution | Western Blot 4 μg/mL, Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 20 μg/mL |
| Applications | Multiplex, Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279873
|
R&D Systems
MAB11623-SP |
25 μg |
£206.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279871
|
R&D Systems
MAB11623-100 |
100 μg |
£486.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Mouse samples. It is validated for Multiplex Immunofluorescence,Western Blot,ImmunohistochemistrySpecifications
| EpCAM/TROP1 | |
| Western Blot 4 μg/mL, Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 20 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
| Mouse myeloma cell line NS0-derived recombinant mouse EPCAM, Accession # Q99JW5 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1085639 | |
| Multiplex, Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q99JW5 | |
| 4072 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title