missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ENPP-7/Alk-SMase Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5160
This item is not returnable.
View return policy
Description
ENPP-7/Alk-SMase Polyclonal specifically detects ENPP-7/Alk-SMase in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| ENPP-7/Alk-SMase | |
| Polyclonal | |
| Unconjugated | |
| NP_001025462 | |
| ENPP7 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ENPP-7/Alk-SMase Ala22-Gln421 Accession # NP_001025462 | |
| 100 μg | |
| Primary | |
| Detects mouse ENPP-7/Alk-SMase in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human ENPP-7 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Alkaline sphingomyelin phosphodiesterase, alkaline sphingomyelinase, ALK-SMase, EC 3.1.4.12, ectonucleotide pyrophosphatase/phosphodiesterase 7, ectonucleotide pyrophosphatase/phosphodiesterase family member 7, E-NPP 7, ENPP7, Intestinal alkaline sphingomyelinase, MGC50179, NPP7, NPP-7 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 339221 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction