missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Endothelin-1 ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
Description
Mouse Endothelin-1 quantitates mouse Endothelin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Endothelin-1.
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Accession Number | ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; Et1; ET-1; HDLCQ7; PPET1; PPET-1; preproendothelin; preproendothelin-1; preproET; QME |
| Assay Range | 4.5 to 1,000 pg/mL |
| Assay Sensitivity | 4.5 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 13614 |
| Gene Symbol | EDN1 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction