missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse EG-VEGF Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
Description
Recombinant Murine EG-VEGF is a 9.6 kDa protein consisting of 86 amino acid residues, including ten cysteine residues that potentially form five pairs of intra-molecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.
Specifications
Specifications
| Accession Number | Q14A28 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 246691 |
| Molecular Weight (g/mol) | 9.6 kDa |
| Name | Mouse EG-VEGF |
| Quantity | 100 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction