missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse E-Cadherin PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
£237.00 - £431.00
Specifications
| Antigen | E-Cadherin |
|---|---|
| Clone | 114420 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18707269
![]() |
R&D Systems
FAB7481P-025 |
25 Tests |
£237.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18797786
![]() |
R&D Systems
FAB7481P-100 |
100 Tests |
£431.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Mouse samples. It is validated for Flow Cytometry.Specifications
| E-Cadherin | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin | |
| CDH1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse E-Cadherin in direct ELISAs and Western blots. |
| 114420 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| Mouse | |
| P09803 | |
| 999 | |
| Mouse myeloma cell line NS0-derived recombinant mouse E-Cadherin Asp157-Val709 (predicted) Accession # P09803 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title