missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse E-Cadherin Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB7481G-100UG
This item is not returnable.
View return policy
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Mouse samples. It is validated for Flow Cytometry.
Specifications
| E-Cadherin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P09803 | |
| CDH1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse E-Cadherin Asp157-Val709 (predicted) Accession # P09803 | |
| RUO | |
| 999 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 114420 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse E-Cadherin in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction