Learn More
Mouse DNAM-1/CD226 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems FAB4436V-100UG
Product Code. 18716579
  • £483.00 / 100µg
Description
DNAM-1/CD226 Monoclonal specifically detects DNAM-1/CD226 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNAM-1/CD226
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q8K4F0
CD226
Mouse myeloma cell line NS0-derived recombinant mouse DNAM-1/CD226 Met1-Pro254 Accession # Q8K4F0
RUO
10666
Mouse
IgG2b
Flow Cytometry
838216
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1
Rat
100 μg
Primary
Detects mouse DNAM-1/CD226 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DNAM-1/CD226 is observed.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
For Research Use Only (RUO)