missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Desmocollin-3 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB7265
This item is not returnable.
View return policy
Description
Desmocollin-3 Monoclonal specifically detects Desmocollin-3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Desmocollin-3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P55850 | |
| DSC3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Desmocollin-3 Arg136-Leu695 Accession # P55850 | |
| 100 μg | |
| Primary | |
| Detects mouse Desmocollin-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 20% cross-reactivity with recombinant human Desmocollin-3 is observed and no cross-reactivity with recombinant mouse (rm) Desmocollin-1 or rmDesmocollin-2 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 765129 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cadherin family member 3, CDHF3, CDHF3HT-CP, desmocollin 3, Desmocollin3, desmocollin-4, DSC, DSC1, DSC2, DSC3, DSC4, DSC4desmocollin-3, HT-CP | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1825 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction