missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Desert Hedgehog/Dhh N-Terminus Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF733
This item is not returnable.
View return policy
Description
Desert Hedgehog/Dhh Polyclonal specifically detects Desert Hedgehog/Dhh in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Desert Hedgehog/Dhh | |
| Polyclonal | |
| Unconjugated | |
| Q61488 | |
| DHH | |
| E. coli-derived recombinant mouse Desert Hedgehog/Dhh aa 23-198 Accession # Q61488 | |
| 100 μg | |
| Primary | |
| Detects mouse Desert Hedgehog/Dhh N-Terminus in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse (rm) Shh N-terminal peptide (aa 25-198) is observed. Additionally, in direct ELISAs, no cross-reactivity with rmDhh C-terminal peptide (aa 199-396) and rmIhh C-terminal peptide (aa 241-449) is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, DHH, HHG-3MGC35145 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 50846 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction