missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Dectin-1/CLEC7A Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1756-100
This item is not returnable.
View return policy
Description
Dectin-1/CLEC7A Monoclonal specifically detects Dectin-1/CLEC7A in Mouse samples. It is validated for Western Blot.
Specifications
| Dectin-1/CLEC7A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q6QLQ4 | |
| CLEC7A | |
| Mouse myeloma cell line NS0-derived recombinant mouse Dectin-1 Phe69-Leu244 Accession # Q6QLQ4 | |
| 100 μg | |
| Primary | |
| Detects mouse Dectin-1 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Dectin-1, rmDectin-2 alpha , or rhDLEC. | |
| Mouse | |
| Supernatant |
| Western Blot | |
| 218838 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Beta-glucan receptor, BGR, CD369, CLEC7A, CLECSF12, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, Dectin1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64581 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction