Learn More
Mouse DcTRAILR1/TNFRSF23 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand:  R&D Systems AF2378
 View more versions of this product
Product Code. 18068631
  • £440.00 / 100µg
Description
DcTRAILR1/TNFRSF23 Polyclonal specifically detects DcTRAILR1/TNFRSF23 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DcTRAILR1/TNFRSF23
Polyclonal
Unconjugated
Q9ER63
Goat
Affinity Purified
RUO
8718
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Tnfrsf23
Mouse myeloma cell line NS0-derived recombinant mouse DcTRAIL R1/TNFRSF23 Met30-Ser156 Accession # Q9ER63
100 μg
Primary
Detects mouse DcTRAIL R1/TNFRSF23 in direct ELISAs and Western blots. In Western blots, approximately 30% cross-reactivity with recombinant mouse (rm) DcTRAIL R2 is observed and less than 1% cross-reactivity with rmTRAIL R2, recombinant human (rh) TRAIL R3, and rhTRAIL R4 is observed.