missing translation for 'onlineSavingsMsg'
Learn More

Mouse DAN Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18798488
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18798488 25 μg 25µg
18139080 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18798488 Supplier R&D Systems Supplier No. MAB7552SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

DAN/NBL1 Monoclonal specifically detects DAN/NBL1 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAN/NBL1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 123224
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q61477
Gene Alias DAN domain family member 1, DAND1, MGC8972, NBL1, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN
Gene Symbols NBL1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse DAN Ala17-Asp178 Accession # Q61477
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4681
Test Specificity Detects mouse DAN in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) DAN, rhCOCO, recombinant mouse (rm) BMP-4, rmGremlin, or recombinant chicken (rch) Caronte is observed. In Western blots, approximately 10% cross-reactivity with rhDAN, rhCOCO, and rchCaronte is observed and no cross reactivity with rmGremlin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.