missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cystatin/Stefin Homolog Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £452.00
Specifications
| Antigen | Cystatin/Stefin Homolog |
|---|---|
| Clone | 263423 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746098
![]() |
R&D Systems
MAB1710-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18140761
|
R&D Systems
MAB1710 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cystatin/Stefin Homolog Monoclonal specifically detects Cystatin/Stefin Homolog in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Cystatin/Stefin Homolog | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P56567 | |
| 1475 | |
| E. coli-derived recombinant mouse Cystatin/Stefin Homolog Ile2-Phe97 Accession # P56567 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Cystatin/Stefin Homolog in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Cystatin D, F, S, SA, SN, recombinant mouse (rm) Cystatin A, B, C, E/M, rmFetuin A, rhFetuin B, rmHPRG, rmKininogen, or rhKininostatin is observed. |
| 263423 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AREI, Cystatin H, PSS4, STF1, STFA | |
| Csta1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title