missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cystatin E/M Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems
Additional Details : Weight : 0.00970kg
Description
Cystatin E/M/CST6 Monoclonal specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
Cystatin E/M/CST6 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 7 ug/mL | |
NP_082899 | |
CST6 | |
Mouse myeloma cell line NS0-derived recombinant mouse Cystatin E/M Glu29-Val149 Accession # NP_082899 | |
500 μg | |
Primary | |
Detects mouse Cystatin E/M in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Cystatin B, S, E/M, SA, S/N, recombinant mouse (rm) Cystatin A, C, D, rhFetuin A, B, rmHPRG, or rhKininogen is observed. | |
Mouse | |
Purified |
Western Blot, Immunoprecipitation, Neutralization | |
216820 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CST6, cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1474 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
For Research Use Only