missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCR3 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £432.00
Specifications
| Antigen | CXCR3 |
|---|---|
| Clone | 220803 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713688
![]() |
R&D Systems
FAB1685P-025 |
25 Tests |
£168.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18757886
![]() |
R&D Systems
FAB1685P-100 |
100 Tests |
£432.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR3 Monoclonal specifically detects CXCR3 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CXCR3 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Mouse | |
| CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
| CXCR3 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse CXCR3. |
| 220803 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| AAC40163 | |
| 2833 | |
| CHO Chinese hamster ovary cell line transfected with mouse CXCR3 Met1-Leu367 Accession # AAC40163 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title