missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCR1/IL-8RA PE-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB8628P-025
This item is not returnable.
View return policy
Description
CXCR1/IL-8RA Monoclonal specifically detects CXCR1/IL-8RA in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CXCR1/IL-8RA | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 RA, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RA, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3577 | |
| Mouse | |
| Supernatant |
| Flow Cytometry | |
| 1122A | |
| Flow Cytometry 10 uL/10^6 cells | |
| NP_839972 | |
| CXCR1 | |
| A peptide corresponding to the N-terminal amino acid sequence of mouse CXCR1 Accession # NP_839972 | |
| 25 Tests | |
| Primary | |
| Detects mouse CXCR1 transfectants but not CXCR2 transfectants in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction