missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL7/Thymus Chemokine-1 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB1091
This item is not returnable.
View return policy
Description
CXCL7/Thymus Chemokine-1 Monoclonal specifically detects CXCL7/Thymus Chemokine-1 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| CXCL7/Thymus Chemokine-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 2-8 ug/mL | |
| Q9EQI5 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5473 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Neutralization | |
| 159742 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Ppbp | |
| E. coli-derived recombinant mouse CXCL7/Thymus Chemokine-1 Lys40-Tyr113 Accession # Q9EQI5 | |
| 500 μg | |
| Primary | |
| Detects mouse CXCL7/Thymus Chemokine-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CXCL1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, 16, recombinant mouse (rm) CXCL1, 4, 6, 9, 10, 11, 12, 13, 15, 16, recombinant rat (rr) CXCL1, 2, 3, 5, recombinant porcine CXCL8, or recombinant cotton rat CXCL10 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction