missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL4/PF4 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB595-SP
This item is not returnable.
View return policy
Description
CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| CXCL4/PF4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5196 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, ELISA | |
| 140910 | |
| Western Blot 2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9Z126 | |
| PF4 | |
| E. coli-derived recombinant mouse CXCL4/PF4 Val30-Ser105 Accession # Q9Z126 | |
| 25 μg | |
| Primary | |
| Detects mouse CXCL4/PF4 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react or interfere recombinant human (rh) CXCL/PF4, recombinant mouse (rm) CXCL9/MIG, rmGM-CSF, rmCXCL11/I-TAC, rhCXCL8/IL-8, rmIL-5, or rhCXCL10/IP-10. In Western blots, no cross-reactivity was observed with rhGCP-2, rmGCP-2, rhGRO alpha , rhGRO beta , rhGRO gamma , rhMIP-1 alpha , rmMIP-1 alpha , rmMIP-1 beta , rhMIP-1 delta , rmMIP-1 gamma , rmMIP-2, rhMIP-3 alpha , rmMIP-3 alpha , rrMIP-3 alpha , rhMIP-3 beta , or rmMIP-3 beta , | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction