missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CXCL2/MIP-2 Quantikine ELISA Kit

Product Code. p-4753790
Click to view available options
Quantity:
2 Plates
6 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18701964

Brand: R&D Systems™ MM200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse CXCL2/MIP-2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CXCL2/MIP-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CXCL2/GRO beta/MIP-2/CINC-3 in cell culture supernates (50 ul), serum (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 1.5pg/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and Recombinant Mouse MIP-2
Sufficient For 2 x 96-well Plates
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 20310
Quantity 2 Plates
Sample Volume Cell Culture Supernates (50uL),Serum (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym chemokine (C-X-C motif) ligand 2, CINC-2 A, CINC3, CINC-3, C-X-C Motif Chemokine 2, GRO beta, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MGSA-beta, MIP2, MIP-2, MIP-2 A, MIP2A, MIP2-alpha, SCYB2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.