missing translation for 'onlineSavingsMsg'
Learn More

Mouse CXCL2/GRO beta/MIP-2/CINC-3 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

£181.00 - £1795.00

Specifications

Antigen CXCL2/GRO beta/MIP-2/CINC-3
Clone 40605
Dilution Western Blot 1 ug/mL, Neutralization 1-4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18733369
GREENER_CHOICE
View Documents
R&D Systems
MAB452-SP
25 μg
£181.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18796409
GREENER_CHOICE
View Documents
R&D Systems
MAB452-100
100 μg
£456.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18782539
GREENER_CHOICE
View Documents
R&D Systems
MAB452-500
500 μg
£1795.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CXCL2/GRO beta/MIP-2/CINC-3 Monoclonal specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

CXCL2/GRO beta/MIP-2/CINC-3
Western Blot 1 ug/mL, Neutralization 1-4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Rat
Mouse
P10889
2920
E. coli-derived recombinant mouse CXCL2/GRO beta/MIP-2/CINC-3 Ala28-Asn100 Accession # P10889
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse CXCL2 in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant human (rh) CXCL2/GRO beta , rhCXCL3/GRO gamma , or recombinant mouse CXCL1/KC.
40605
Western Blot, Neutralization, ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
chemokine (C-X-C motif) ligand 2, CINC-2a, CINC3, CINC-3, C-X-C Motif Chemokine 2, GRO beta, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MGSA-beta, MIP2, MIP-2, MIP-2a, MIP2A, MIP2-alpha, SCYB2
CXCL2
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.