missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL17/VCC-1 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF4270
Additional Details : Weight : 0.00970kg
Description
CXCL17/VCC-1 Polyclonal specifically detects CXCL17/VCC-1 in Mouse samples. It is validated for Western Blot.Specifications
CXCL17/VCC-1 | |
Polyclonal | |
Unconjugated | |
Q5UW37 | |
CXCL17 | |
E. coli-derived recombinant mouse CXCL17/VCC-1 Ser31-Leu119 Accession # Q5UW37 | |
100 μg | |
Primary | |
Detects mouse CXCL17/VCC-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human CXCL17/VCC-1 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Chemokine (C-X-C Motif) Ligand 17, C-X-C Motif Chemokine 17, CXCL17, Dcip1, Dendritic Cell And Monocyte Chemokine-Like Protein, DMC, UNQ473, VCC1, VCC-1, VEGF Coregulated Chemokine 1, VEGF Co-Regulated Chemokine 1 | |
Sheep | |
Affinity Purified | |
RUO | |
284340 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only