missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL17/VCC-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4270-SP
This item is not returnable.
View return policy
Description
CXCL17/VCC-1 Monoclonal specifically detects CXCL17/VCC-1 in Mouse samples. It is validated for Western Blot.
Specifications
| CXCL17/VCC-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q5UW37 | |
| CXCL17 | |
| E. coli-derived recombinant mouse CXCL17/VCC-1 Ser31-Leu119 Accession # Q5UW37 | |
| 25 μg | |
| Primary | |
| Detects recombinant mouse (rm) CXCL17/VCC-1 in direct ELISAs and Western blots. In direct ELISAs, 50% cross-reactivity with recombinant human CXCL17 is observed/VCC-1. | |
| Mouse | |
| Purified |
| Western Blot | |
| 510614 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Chemokine (C-X-C Motif) Ligand 17, C-X-C Motif Chemokine 17, CXCL17, Dcip1, Dendritic Cell And Monocyte Chemokine-Like Protein, DMC, UNQ473, VCC1, VCC-1, VEGF Coregulated Chemokine 1, VEGF Co-Regulated Chemokine 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 284340 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction