missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse CXCL16 Recombinant Protein, PeproTech®

Product Code. 17811103
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
5µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17811103 25 μg 25µg
17860613 100 μg 100µg
17880343 1 mg 1mg
17830543 250 μg 250µg
17870913 500 μg 500µg
17890613 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17811103 Supplier Gibco™ Supplier No. 2502825UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine CXCL16 is a 9.9 kDa protein containing 88 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL16 is a member of the CXC chemokine family and signals through the CXCR6 receptor. CXCL16 may play a role in attracting lymphocyte subsets during inflammation and may facilitate certain immune responses. The chemokine domain of CXCL16 contains six cysteine residues, including the four highly conserved cysteine residues characteristic of CXC chemokines. The CXCL16 gene codes for a 273 amino acid polypeptide, which includes a 29 amino acid cytoplasmic domain and transmembrane sequence containing approximately 20 amino acids. The extracellular portion of CXCL16 contains a chemokines domain and an extended C-terminal mucin-like stalk sequence. The extracellular domain contains 89 amino acid residues (86 a.a. residues for the murine homolog).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8BSU2
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 66102
Molecular Weight (g/mol) 9.9 kDa
Name Mouse CXCL16
Quantity 25 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 0910001K24Rik; AV290116; b2b498Clo; BB024863; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 16; CXC; Cxc chemokine ligand 16; CXC motif chemokine; C-X-C motif chemokine 16; C-X-C motif chemokine ligand 16; CXCL; Cxcl16; CXCL16v1; CXCL16v2; CXCLG16; Mutant line 498; scavenger receptor for phosphatidylserine and oxidized low density lipoprotein; SCYB16; Small-inducible cytokine B16; Srpsox; SR-PSOX; SR-PSOX/CXCL16; Transmembrane chemokine CXCL16; UNQ2759/PRO6714; zinc finger, MYND-type containing 15; Zmynd15
Common Name CXCL16
Gene Symbol Cxcl16
Biological Activity Determined by its ability to chemoattract murine lymphocytes using a concentration of 20-1000 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence NQGSVAGSCS CDRTISSGTQ IPQGTLDHIR KYLKAFHRCP FFIRFQLQSK SVCGGSQDQW VRELVDCFER KECGTGHGKS FHHQKHLP
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.