missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL16 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
£170.00 - £1580.00
Specifications
| Antigen | CXCL16 |
|---|---|
| Clone | 142417 |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.15-0.6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18767408
![]() |
R&D Systems
MAB503-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18789397
![]() |
R&D Systems
MAB503-100 |
100 μg |
£389.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18775579
![]() |
R&D Systems
MAB503-500 |
500 μg |
£1580.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCL16 Monoclonal specifically detects CXCL16 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CXCL16 | |
| Western Blot 1 ug/mL, Neutralization 0.15-0.6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q8BSU2 | |
| 58191 | |
| E. coli-derived recombinant mouse CXCL16 Asn27-Pro114 Accession # Q8BSU2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CXCL16 in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, recombinant mouse CXCL1, 2, 6, 9, 10, 11, 12, 13, 14, 15, recombinant rat CXCL1, 2, 3, 5, or recombinant porcine CXCL8 is observed. |
| 142417 | |
| Western Blot, Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCL16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16 | |
| CXCL16 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title