missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL13/BLC/BCA-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB470-SP
This item is not returnable.
View return policy
Description
CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CXCL13/BLC/BCA-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10563 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization, ELISA | |
| 143614 | |
| Neutralization 12-30 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O55038 | |
| CXCL13 | |
| E. coli-derived recombinant mouse CXCL13 Ile22-Ala109 Accession # O55038 | |
| 25 μg | |
| Primary | |
| Detects mouse CXCL13 in ELISAs. In ELISAs, this antibody does not cross-react with recombinant human (rh) CXCL13, rmCXCL11/I-TAC, rmCXCL9/MIG, or rm CXCL12/SDF-1 alpha . | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction